Skip to Content
Merck
All Photos(1)

Key Documents

707058

Sigma-Aldrich

7-Fluoro-5-iodoindole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€84.00

€84.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€84.00

About This Item

Empirical Formula (Hill Notation):
C8H5FIN
CAS Number:
Molecular Weight:
261.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€84.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

functional group

fluoro
iodo

storage temp.

2-8°C

SMILES string

Fc1cc(I)cc2cc[nH]c12

InChI

1S/C8H5FIN/c9-7-4-6(10)3-5-1-2-11-8(5)7/h1-4,11H

InChI key

BRUNWOSWRZMWNY-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service