Skip to Content
Merck
All Photos(2)

Key Documents

706655

Sigma-Aldrich

2-Thiophenesulfonamide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO2S2
CAS Number:
Molecular Weight:
163.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%
To inquire about this product 706655, please contact your local Merck office or dealer. Contact Technical Service

Assay

96%

form

solid

mp

137-141 °C

SMILES string

NS(=O)(=O)c1cccs1

InChI

1S/C4H5NO2S2/c5-9(6,7)4-2-1-3-8-4/h1-3H,(H2,5,6,7)

InChI key

KTFDYVNEGTXQCV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuhal Alım et al.
Pharmacological reports : PR, 72(6), 1738-1748 (2020-08-05)
Thiophene(s) are an important group in therapeutic applications, and sulfonamides are the most important class of carbonic anhydrase (CA) inhibitors. In this study, inhibition effects of some thiophene-based sulfonamides on human erythrocytes carbonic anhydrase I and II isoenzymes (hCA-I and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service