Skip to Content
Merck
All Photos(1)

Key Documents

702730

Sigma-Aldrich

tert-Butylchlorodiphenylsilane solution

1 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClSi
CAS Number:
Molecular Weight:
274.86
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1 M in methylene chloride

bp

40 °C

density

1.249 g/mL at 25 °C

SMILES string

CC(C)(C)[Si](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C16H19ClSi/c1-16(2,3)18(17,14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3

InChI key

MHYGQXWCZAYSLJ-UHFFFAOYSA-N

Application

tert-Butylchlorodiphenylsilane can be used to prepare 1-benzyloxy-3-(tert-butyldiphenylsilyloxy)propan-2-ol, a key intermediate for the synthesis of mono-O-protected pyrimidine acyclic nucleosides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new synthetic route towards the mono-O-protected anti-conformationally constrained pyrimidine acyclic nucleoside
Liu C, et al.
Chemical & Pharmaceutical Bulletin, 50(8), 1028-1030 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service