Skip to Content
Merck
All Photos(1)

Key Documents

693782

Sigma-Aldrich

Neodymium-iron-boron alloy 30/150

disc, size 13 mm × 6 mm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

disc

composition

Aluminum, 0.8 wt. %
Boron, 1.1 wt. %
Dysprosium, 3.6 wt. %
Iron, 65 wt. %
Neodymium, 29 wt. %
Niobium, 0.5 wt. %

size

13 mm × 6 mm

SMILES string

[B].[Fe].[Nd]

InChI

1S/B.Fe.Nd

InChI key

QJVKUMXDEUEQLH-UHFFFAOYSA-N

Other Notes

Magnetic parameters/properties:
Maximum Energy Product (BH): 247 kJ/m3 (31 MGsOe)
Residual Induction (Br): 1.13 T (11.3 kGs)
Coercive Force (Hc): 844 kA/m (10.6 kOe)
Intrinsic Coercive Force (Hci): 1595 kA/m (20.0 kOe)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Self-heat. 1 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service