693502
Gadolinium-silicon-germanium alloy
Gd5Si0.5Ge3.5, ≥99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Gd5Ge2Si2
Molecular Weight:
987.70
UNSPSC Code:
12352300
Assay
≥99%
form
powder and chunks
composition
Gadolinium, 74.4 wt. %
Germanium, 24.2 wt. %
Silicon, 1.4 wt. %
mp
1313 °C
Application
Physical properties
Curie Temperature: ~70 K
Legal Information
Sold under license to US Patents 5,887,449 and 5,743,095 for use in magnetic refrigeration.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 3
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service