Skip to Content
Merck
All Photos(1)

Key Documents

691283

Sigma-Aldrich

Butyl cyanoacetate

Arxada quality, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CO2(CH2)3CH3
CAS Number:
Molecular Weight:
141.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.10% water

color

APHA: ≤60

refractive index

n20/D 1.425 (lit.)

bp

115 °C/15 mmHg (lit.)

density

0.998-1.002 g/mL at 20 °C
0.993 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CC#N

InChI

1S/C7H11NO2/c1-2-3-6-10-7(9)4-5-8/h2-4,6H2,1H3

InChI key

DJACTCNGCHPGOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service