Skip to Content
Merck
All Photos(1)

Key Documents

690848

Sigma-Aldrich

N,N-Dimethylacetoacetamide solution

Arxada quality, 79.0-83.0% (calculated)

Synonym(s):

N,N-Dimethyl-3-oxobutyramide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CON(CH3)2
CAS Number:
Molecular Weight:
129.16
Beilstein:
1755038
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

79.0-83.0% (calculated)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

impurities

≤0.05% dimethylamine

color

APHA: ≤350

SMILES string

CN(C)C(=O)CC(C)=O

InChI

1S/C6H11NO2/c1-5(8)4-6(9)7(2)3/h4H2,1-3H3

InChI key

YPEWWOUWRRQBAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bharathi Avula et al.
Journal of AOAC International, 98(3), 757-759 (2015-06-19)
The central nervous system stimulant 1,3-dimethylamylamine (DMAA) has been found in preworkout products and dietary supplements. A fast direct analysis in real time-quadrupole time of flight-MS method was used for identification of DMAA in dietary supplements and to determine if
Kyung Tai Lee et al.
Foot & ankle international, 35(12), 1329-1333 (2014-09-23)
Hallux valgus is speculated to increase the load on the second metatarsophalangeal (MTP) joint, possibly inducing degenerative osteoarthritis. In addition, the severity of arthritis may be correlated with the severity of hallux valgus. This study evaluated the association of arthritis
Sonia D'Souza et al.
Biomaterials, 35(35), 9447-9458 (2014-08-26)
The field of polymer-based membrane engineering has expanded since we first demonstrated the reaction of N-hydroxysuccinimide ester-terminated polymers with cells and tissues almost two decades ago. One remaining obstacle, especially for conjugation of polymers to cells, has been that exquisite
Marek Janko et al.
Biomacromolecules, 16(7), 2179-2187 (2015-06-24)
The properties of paper sheets can be tuned by adjusting the surface or bulk chemistry using functional polymers that are applied during (online) or after (offline) papermaking processes. In particular, polymers are widely used to enhance the mechanical strength of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service