Skip to Content
Merck
All Photos(1)

Key Documents

688525

Sigma-Aldrich

(S,S)-N-Boc-2,5-dimethylpyrrolidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO2
CAS Number:
Molecular Weight:
199.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

optical activity

[α]20/D +45.0°, c = 1 in chloroform

refractive index

n20/D 1.444

density

0.930 g/mL at 25 °C

SMILES string

C[C@H]1CC[C@H](C)N1C(=O)OC(C)(C)C

InChI

1S/C11H21NO2/c1-8-6-7-9(2)12(8)10(13)14-11(3,4)5/h8-9H,6-7H2,1-5H3/t8-,9-/m0/s1

InChI key

FZSBLLYQAULJDU-IUCAKERBSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

187.0 °F - closed cup

Flash Point(C)

86.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service