684481
[6,6]-Phenyl-C61 butyric acid octyl ester
≥99%
Synonym(s):
1-(3-Octoxycarbonylpropyl)-1-phenyl-[6.6]C61, PCBO, [60]PCB-C8
About This Item
Recommended Products
description
functionalized fullerene
Assay
≥99%
form
powder
SMILES string
CCCCCCCCOC(=O)CCCC2(c1ccccc1)[C]3=4c5c6c7c8c9c%10c(c%11c%12c3c%13c5c%14c%15c6c%16c7c%17c9c%18c%19c%10c%20c%11c%21c%12c%22c%13c%23c%14c%24c%15c%25c%16c%26c%17c%18c%27c%28c%19c%20c%29c%21c%30c%22c%23c%31c%24c%32c%25c%26c%27c%33c%28c%29c%30c%31c%32%33)[C]2=48
InChI
1S/C59H38O2/c1-2-3-4-5-6-10-26-61-38(60)14-11-25-57(27-12-8-7-9-13-27)58-36-23-24-37-35-22-20-33-31-18-16-29-28-15-17-30-32-19-21-34(36)45-43(32)48-41(30)39(28)47-40(29)42(31)49-44(33)46(35)56(59(37,57)58)54-52(49)50(47)51(48)53(54)55(45)58/h7-9,12-13,15-24H,2-6,10-11,14,25-26H2,1H3/t57-,58-,59+
InChI key
BRVSNRNVRFLFLL-PWHDIVFGSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.
PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.
Find various photovoltaic and bioscience-based applications of fullerenes.
Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service