Skip to Content
Merck
All Photos(1)

Key Documents

682004

Sigma-Aldrich

N-Benzyl-N-(2-quinolinylmethyl)formamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O
CAS Number:
Molecular Weight:
276.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

57-62 °C

SMILES string

[H]C(=O)N(Cc1ccccc1)Cc2ccc3ccccc3n2

InChI

1S/C18H16N2O/c21-14-20(12-15-6-2-1-3-7-15)13-17-11-10-16-8-4-5-9-18(16)19-17/h1-11,14H,12-13H2

InChI key

UCIKFROUISXGMV-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service