Skip to Content
Merck
All Photos(1)

Key Documents

674230

Sigma-Aldrich

11-Mercaptoundecyl trifluoroacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23F3O2S
CAS Number:
Molecular Weight:
300.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.4294

mp

50-51 °C

density

1.059 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)OCCCCCCCCCCCS

InChI

1S/C13H23F3O2S/c14-13(15,16)12(17)18-10-8-6-4-2-1-3-5-7-9-11-19/h19H,1-11H2

InChI key

YOZNBMSWVPXLKM-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service