Skip to Content
Merck
All Photos(1)

Key Documents

656372

Sigma-Aldrich

5-Bromo-2,4-difluoroaniline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(F)2NH2
CAS Number:
Molecular Weight:
208.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

25-29 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1cc(Br)c(F)cc1F

InChI

1S/C6H4BrF2N/c7-3-1-6(10)5(9)2-4(3)8/h1-2H,10H2

InChI key

FQZCUAASVCIWSL-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service