Skip to Content
Merck
All Photos(1)

Key Documents

653950

Sigma-Aldrich

trans-6-Chloro-1-hexen-1-ylboronic acid pinacol ester

96%

Synonym(s):

(E)-6-Chloro-1-hexenylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H22BClO2
CAS Number:
Molecular Weight:
244.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

96%

mp

38-42 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)\C=C\CCCCCl

InChI

1S/C12H22BClO2/c1-11(2)12(3,4)16-13(15-11)9-7-5-6-8-10-14/h7,9H,5-6,8,10H2,1-4H3/b9-7+

InChI key

FNDLLXXDZTUGLB-VQHVLOKHSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service