All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4HCl3O2S2
CAS Number:
Molecular Weight:
251.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
54-58 °C (lit.)
storage temp.
2-8°C
SMILES string
Clc1cc(sc1Cl)S(Cl)(=O)=O
InChI
1S/C4HCl3O2S2/c5-2-1-3(10-4(2)6)11(7,8)9/h1H
InChI key
IVTWLTRKVRJPNG-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service