Skip to Content
Merck
All Photos(1)

Documents

648477

Sigma-Aldrich

Methyl 5-(4-cyanophenyl)furan-2-carboxylate

96%

Synonym(s):

5-(4-cyanophenyl)furan-2-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO3
CAS Number:
Molecular Weight:
227.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

154-158 °C (lit.)

SMILES string

COC(=O)c1ccc(o1)-c2ccc(cc2)C#N

InChI

1S/C13H9NO3/c1-16-13(15)12-7-6-11(17-12)10-4-2-9(8-14)3-5-10/h2-7H,1H3

InChI key

KXWHTAUZNSVTPO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service