Skip to Content
Merck
All Photos(1)

Key Documents

642606

Sigma-Aldrich

trans-3-Phenyl-1-propen-1-ylboronic acid

≥95%

Synonym(s):

trans-3-phenylpropen-1-yl-boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€151.00
5 G
€519.00

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€151.00
5 G
€519.00

About This Item

Empirical Formula (Hill Notation):
C9H11BO2
CAS Number:
Molecular Weight:
161.99
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€151.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95%

form

solid

functional group

phenyl

SMILES string

[H]\C(Cc1ccccc1)=C(\[H])B(O)O

InChI

1S/C9H11BO2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-6,8,11-12H,7H2/b8-4+

InChI key

GMGWFDHLFMBIDS-XBXARRHUSA-N

Application

Reactant involved in:
  • Carbonylative arylation of allenols
  • Oxidative cross-coupling with (trifluoromethyl)trimethylsilane
  • Suzuki-Miyaura coupling reactions
  • three-component reductive coupling with alkynes
  • Enantioselective conjugation to vinyl 2-pyridylsulfones

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ioan-Adrian Stoian et al.
Biosensors & bioelectronics, 155, 112098-112098 (2020-02-25)
A highly selective and sensitive molecularly imprinted polymer (MIP)-based electrochemical sensor was fabricated for the determination of azithromycin, a broad-spectrum macrolide antibiotic, from various biological samples (urine, tears, plasma). The reversible boronate ester bond-mediated, thin (~75 nm) MIP-based biomimetic recognition layer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service