Skip to Content
Merck
All Photos(2)

Documents

632155

Sigma-Aldrich

2-Chloro-3-pyridinecarboxaldehyde

97%

Synonym(s):

Chloro-2-formyl-3-pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO
CAS Number:
Molecular Weight:
141.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

50-54 °C (lit.)

SMILES string

[H]C(=O)c1cccnc1Cl

InChI

1S/C6H4ClNO/c7-6-5(4-9)2-1-3-8-6/h1-4H

InChI key

KHPAGGHFIDLUMB-UHFFFAOYSA-N

Application

Starting material for the preparation of 5-azaindoles by the Hemetsberger-Knittel reaction involving the thermal decomposition of alkenyl azides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 2751-2751 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service