Skip to Content
Merck
All Photos(1)

Key Documents

631655

Sigma-Aldrich

2,2′-Bis(methoxymethoxy)-1,1′-binaphthalene

97%

Synonym(s):

2,2′-Bis(methoxymethoxy)-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2OC10H6)2
CAS Number:
Molecular Weight:
374.43
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reagent type: ligand

mp

92-96 °C (lit.)

SMILES string

COCOc1ccc2ccccc2c1-c3c(OCOC)ccc4ccccc34

InChI

1S/C24H22O4/c1-25-15-27-21-13-11-17-7-3-5-9-19(17)23(21)24-20-10-6-4-8-18(20)12-14-22(24)28-16-26-2/h3-14H,15-16H2,1-2H3

InChI key

YIAQRNNJNMLGTP-UHFFFAOYSA-N

General description

2,2′-Bis(methoxymethoxy)-1,1′-binaphthalene is an aryl alkyl ether that can undergo nickel catalyzed cross-coupling reaction with aryl Grignard reagent to form the aryl-aryl cross-coupling product.[1]

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-Coupling of Aryl/Alkenyl Ethers with Aryl Grignard Reagents through Nickel-Catalyzed C? O Activation.
Xie LG and Wang ZX.
Chemistry?A European Journal , 17(18), 4972-4975 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service