Skip to Content
Merck
All Photos(1)

Key Documents

630357

Sigma-Aldrich

2-Acetoxyphenylzinc iodide

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4ZnI
Molecular Weight:
327.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

0.5 M in THF

impurities

1-3% inorganic chloride

bp

65 °C

density

1.033 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccccc1[Zn]I

InChI

1S/C8H7O2.HI.Zn/c1-7(9)10-8-5-3-2-4-6-8;;/h2-5H,1H3;1H;/q;;+1/p-1

InChI key

DUYNRVQSBJWXDT-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service