Skip to Content
Merck
All Photos(1)

Documents

608882

Sigma-Aldrich

Ammonium-15N acetate

40-70 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO215NH4
CAS Number:
Molecular Weight:
78.08
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:

isotopic purity

40-70 atom % 15N

mol wt

77.63 by atom % calculation (at 55 atom % 15N)

mp

110-112 °C (dec.) (lit.)

mass shift

M+1

SMILES string

CC([O-])=O.[H][15N+]([H])([H])[H]

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3/i;1+1

InChI key

USFZMSVCRYTOJT-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service