Skip to Content
Merck
All Photos(1)

Key Documents

597600

Sigma-Aldrich

4-Amino-2-chloropyrimidine-5-carbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClN4
CAS Number:
Molecular Weight:
154.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

200 °C (dec.) (lit.)

functional group

chloro
nitrile

SMILES string

Nc1nc(Cl)ncc1C#N

InChI

1S/C5H3ClN4/c6-5-9-2-3(1-7)4(8)10-5/h2H,(H2,8,9,10)

InChI key

WDHFCSOENXEMRC-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Pevec
Acta chimica Slovenica, 62(2), 297-303 (2015-06-19)
The synthesis and spectroscopic characterization of some pyrimidinium hexafluoridosilicate salts with the formula (RH)2[SiF]6 (where R = 2-aminopyrimidine (1), cytosine (2) and 5-cyanocytosine (3)) is obtained in the reaction of SiO2 with hydrofluoric acid solution and subsequent addition of 2-aminopyrimidine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service