591327
Poly(1,1,1,3,3,3-hexafluoroisopropyl methacrylate)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
refractive index
n20/D 1.39 (lit.)
transition temp
Tg (DSC) 56 °C(lit.)
Tg 56 °C(lit.)
solubility
DMSO: soluble
NMP: soluble
THF: soluble
Related Categories
Application
Low refractive index, photocrosslinkable polymer for cladding layer in optical waveguides. Transparent, low surface tension polymer.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service