Skip to Content
Merck
All Photos(1)

Documents

588237

Sigma-Aldrich

Benzyl bromide-(phenyl-13C6)

99 atom % 13C

Synonym(s):

α-Bromotoluene-(phenyl-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5CH2Br
CAS Number:
Molecular Weight:
176.99
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

198-199 °C (lit.)

mp

-3--1 °C (lit.)

density

1.488 g/mL at 25 °C

mass shift

M+6

SMILES string

BrC[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1+1,2+1,3+1,4+1,5+1,7+1

InChI key

AGEZXYOZHKGVCM-ZXJNGCBISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service