566233
Poly[(methyl methacrylate)-co-(7-(4-trifluoromethyl)coumarin acrylamide)]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
composition
4-(trifluoromethyl)coumarin-N-acrylamide, ~25 mol %
solubility
chloroform: soluble
toluene: soluble
fluorescence
λex 331 nm; λem 418 nm in chloroform
Application
Fluorescent polymer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service