Skip to Content
Merck
All Photos(1)

Key Documents

560057

Sigma-Aldrich

9,9-Didodecyl-2,7-dibromofluorene

97%

Synonym(s):

2,7-Dibromo-9,9-didodecyl-9H -fluorene, 2,7-Dibromo-9,9-didodecylfluorene, 9,9-Didodecyl-2,7-dibromofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H56Br2
CAS Number:
Molecular Weight:
660.65
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

bp

180 °C/0.15 mmHg (lit.)

mp

50-55 °C (lit.)

SMILES string

CCCCCCCCCCCCC1(CCCCCCCCCCCC)c2cc(Br)ccc2-c3ccc(Br)cc13

InChI

1S/C37H56Br2/c1-3-5-7-9-11-13-15-17-19-21-27-37(28-22-20-18-16-14-12-10-8-6-4-2)35-29-31(38)23-25-33(35)34-26-24-32(39)30-36(34)37/h23-26,29-30H,3-22,27-28H2,1-2H3

InChI key

KFOUJVGPGBSIFB-UHFFFAOYSA-N

Application

Intermediate for polymeric light-emitting diodes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service