Skip to Content
Merck
All Photos(3)

Key Documents

557803

Sigma-Aldrich

2,6-Dichloroquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5Cl2N
CAS Number:
Molecular Weight:
198.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

161-164 °C (lit.)

functional group

chloro

SMILES string

Clc1ccc2nc(Cl)ccc2c1

InChI

1S/C9H5Cl2N/c10-7-2-3-8-6(5-7)1-4-9(11)12-8/h1-5H

InChI key

LPDFGLZUUCLXGM-UHFFFAOYSA-N

General description

2,6-Dichloroquinoline can be synthesized from 6-chloroquinoline. It can also be prepared from the corresponding 2-vinylaniline.

Application

2,6-Dichloroquinoline may be used in the preparation of poly(quinoline-2,6-diyl), via electrochemical polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical and chemical preparation of linear. pi.-conjugated poly (quinoline-2, 6-diyl) using nickel complexes and electrochemical properties of the polymer.
Saito N, et al.
Macromolecules, 27(3), 756- 761 (1994)
Poly (1, 5-naphthyridine-2, 6-diyl) Having a Highly Extended and Electron-Withdrawing. pi.-Conjugation System. Preparation, Optical Properties, and Electrochemical Redox Reaction.
Saito N and Yamamoto T.
Macromolecules, 28(12), 4260-4267 (1995)
Byoung Se Lee et al.
The Journal of organic chemistry, 67(22), 7884-7886 (2002-10-26)
2-Vinyl- or heteroaryl-substituted anilines were reacted with diphosgene in acetonitrile solution via a reactive imidoyl moiety to afford the corresponding 2-chloroquinolines. Facile syntheses of nine 2-chloroquinoline derivatives from several anilines and their postulate mechanism is described. The postulate mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service