Skip to Content
Merck
All Photos(1)

Key Documents

552372

Sigma-Aldrich

5-Chloro-1-(4-piperidyl)-2-benzimidazolinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14ClN3O
CAS Number:
Molecular Weight:
251.71
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

228-231 °C (lit.)

SMILES string

Clc1ccc2N(C3CCNCC3)C(=O)Nc2c1

InChI

1S/C12H14ClN3O/c13-8-1-2-11-10(7-8)15-12(17)16(11)9-3-5-14-6-4-9/h1-2,7,9,14H,3-6H2,(H,15,17)

InChI key

DOAYWDKFDPSTSV-UHFFFAOYSA-N

General description

5-Chloro-1-(4-piperidyl)-2-benzimidazolinone is a 2-benzimidazolone derivative. It exhibits anti-nauseant properties.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed efficient synthesis of a 2-benzimidazolone scaffold from 2-nitroaniline and dimethyl carbonate via a hydrosilylation reaction.
Nale DB and Bhanage BM.
Green Chemistry, 17(4), 2480-2486 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service