Skip to Content
Merck
All Photos(1)

Documents

544450

Sigma-Aldrich

4,4′-Biphenyldicarbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4C6H4CN
CAS Number:
Molecular Weight:
204.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

236-240 °C (lit.)

SMILES string

N#Cc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C14H8N2/c15-9-11-1-5-13(6-2-11)14-7-3-12(10-16)4-8-14/h1-8H

InChI key

KAXYYLCSSXFXKR-UHFFFAOYSA-N

General description

4,4′-Biphenyldicarbonitrile (BPCN), also known as 4,4′-dicyanobiphenyl, is a para-disubstituted biphenyl. It can be prepared from benzidine dihydrochloride.3 BPCN belongs to the monoclinic crystal system and P21 space group.

Application

4,4′-Biphenyldicarbonitrile (BPCN, 4,4′-dicyanobiphenyl) may be used in the preparation of:
  • [Ag(BPCN)2]PF6 (PF6=Hexafluorophosphate)
  • 4,4′-diphenacetylbiphenyl
  • triazine-framework-based porous membranes (TFMs)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization of 4,4?-biphenyldicarbonitrile with silver (I) salts: a change in topology concomitant with a change in counterion leading to a ninefold diamondoid network.
Hirsch KA, et al.
Journal of the Chemical Society. Chemical Communications, 21, 2199-2200 (1995)
Bistetracyclones? and ?Bishexaphenylbenzenes.
Ogliaruso MA, et al.
The Journal of Organic Chemistry, 28(10), 2725-2728 (1963)
A superacid-catalyzed synthesis of porous membranes based on triazine frameworks for CO2 separation.
Zhu X, et al.
Journal of the American Chemical Society, 134(25), 10478-10484 (2012)
4, 4?-Dicyanobiphenyl.
Britton D and Young Jr VG.
Acta Crystallographica Section E, Structure Reports Online, 59(11), o1849-o1851 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service