Skip to Content
Merck
All Photos(1)

Key Documents

528471

Sigma-Aldrich

Ethyl trans-4-ethoxycinnamate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CH=CHCO2C2H5
CAS Number:
Molecular Weight:
220.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

38-42 °C (lit.)

SMILES string

[H]\C(=C(\[H])c1ccc(OCC)cc1)C(=O)OCC

InChI

1S/C13H16O3/c1-3-15-12-8-5-11(6-9-12)7-10-13(14)16-4-2/h5-10H,3-4H2,1-2H3/b10-7+

InChI key

XFRWZVXVZIQXFI-JXMROGBWSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service