Skip to Content
Merck
All Photos(1)

Key Documents

528153

Sigma-Aldrich

2,6-Dimethylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
1903544
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

bp

262 °C (lit.)

mp

106-110 °C (lit.)

SMILES string

Cc1ccc2cc(C)ccc2c1

InChI

1S/C12H12/c1-9-3-5-12-8-10(2)4-6-11(12)7-9/h3-8H,1-2H3

InChI key

YGYNBBAUIYTWBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of BP Amoco

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service