Skip to Content
Merck
All Photos(1)

Documents

524093

Sigma-Aldrich

2,3,6-Trifluorophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2F3B(OH)2
CAS Number:
Molecular Weight:
175.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

127-132 °C (lit.)

SMILES string

OB(O)c1c(F)ccc(F)c1F

InChI

1S/C6H4BF3O2/c8-3-1-2-4(9)6(10)5(3)7(11)12/h1-2,11-12H

InChI key

IWPDDRPLEKURGG-UHFFFAOYSA-N

General description

Contains varying amounts of anhydride.

Application

Reactant for:
  • Synthesis of C-6 hydroxy tricyclic sulfone as a γ-secretase inhibitor via Suzuki coupling reaction
  • Palladium catalyzed Suzuki-Miyaura coupling reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service