Skip to Content
Merck
All Photos(1)

Documents

51850

Sigma-Aldrich

Heptatriacontane

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)35CH3
CAS Number:
Molecular Weight:
521.00
Beilstein:
1713110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0%

mp

77-79 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C37H76/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-37H2,1-2H3

InChI key

PXEZIKSRSYGOED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service