Skip to Content
Merck
All Photos(1)

Key Documents

512311

Sigma-Aldrich

2,5-Dichlorophenylboronic acid

Synonym(s):

2,5-Dichlorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3B(OH)2
CAS Number:
Molecular Weight:
190.82
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

150 °C (lit.)

SMILES string

OB(O)c1cc(Cl)ccc1Cl

InChI

1S/C6H5BCl2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,10-11H

InChI key

NNTFPBXQPOQRBT-UHFFFAOYSA-N

Application

Reactant for:
  • Regioselective iodination using silver salts
  • Pd-catalyzed Suzuki-Miyaura coupling
  • Rhodium(I)-catalyzed carbonylative cyclization of alkynes
  • Copper catalyzed coupling reactions

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service