499269
1-Methylbutylzinc bromide solution
0.5 M in THF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2CH(CH3)ZnBr
CAS Number:
Molecular Weight:
216.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
concentration
0.5 M in THF
density
0.961 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CCCC(C)[Zn]Br
InChI
1S/C5H11.BrH.Zn/c1-3-5-4-2;;/h3H,4-5H2,1-2H3;1H;/q;;+1/p-1
InChI key
CIYMRBSCGOXOEW-UHFFFAOYSA-M
Legal Information
Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.0 °F - closed cup
Flash Point(C)
-17.22 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service