481041
Titanium(II) oxide
powder, −325 mesh, 99.9% trace metals basis
Synonym(s):
Titanium monoxide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
TiO
CAS Number:
Molecular Weight:
63.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
deposition grade
Assay
99.9% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: titanium
particle size
−325 mesh
SMILES string
O=[Ti]
InChI
1S/O.Ti
InChI key
OGIDPMRJRNCKJF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Donald J Joseph et al.
PloS one, 14(10), e0223509-e0223509 (2019-10-11)
Many in vivo studies suggest that inhalational anesthetics can accelerate or prevent the progression of neuropathology and cognitive impairments in Alzheimer Disease (AD), but the synaptic mechanisms mediating these ambiguous effects are unclear. Here, we show that repeated exposures of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service