Skip to Content
Merck
All Photos(1)

Documents

479926

Sigma-Aldrich

(S)-(−)-N-Boc-5-benzyl-2-oxomorpholine

99%

Synonym(s):

tert-Butyl (S)-(−)-5-benzyl-2-oxo-4-morpholinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO4
CAS Number:
Molecular Weight:
291.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −14.5°, c = 2.45 in chloroform

mp

98-100 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CC(=O)OC[C@@H]1Cc2ccccc2

InChI

1S/C16H21NO4/c1-16(2,3)21-15(19)17-10-14(18)20-11-13(17)9-12-7-5-4-6-8-12/h4-8,13H,9-11H2,1-3H3/t13-/m0/s1

InChI key

NQZZZVQLJCFCGL-ZDUSSCGKSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service