Skip to Content
Merck
All Photos(1)

Key Documents

47508

Sigma-Aldrich

Fmoc-D-Ala-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO4
CAS Number:
Molecular Weight:
311.33
Beilstein:
8025133
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (HPLC)

optical activity

[α]20/D +18.5±1°, c = 1% in DMF

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

C[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m1/s1

InChI key

QWXZOFZKSQXPDC-LLVKDONJSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Schneider et al.
Nature communications, 9(1), 4396-4396 (2018-10-26)
New chemical inhibitors of protein-protein interactions are needed to propel advances in molecular pharmacology. Peptoids are peptidomimetic oligomers with the capability to inhibit protein-protein interactions by mimicking protein secondary structure motifs. Here we report the in silico design of a
Si Chen et al.
Biomaterials, 117, 92-104 (2016-12-13)
In this work, mitochondria-targeting gold nanostar (AuNS) and anticarcinogen DOX were co-encapsulated in hyaluronic acid (HA) protective shell for tumor-targeting synergistic photothermal/chemo-therapy. Cationic peptide R
Fayçal Touti et al.
Nature chemical biology, 15(4), 410-418 (2019-03-20)
The use of competitive inhibitors to disrupt protein-protein interactions (PPIs) holds great promise for the treatment of disease. However, the discovery of high-affinity inhibitors can be a challenge. Here we report a platform for improving the affinity of peptide-based PPI
Andreas Pech et al.
Nucleic acids research, 45(7), 3997-4005 (2017-02-06)
Biological evolution resulted in a homochiral world in which nucleic acids consist exclusively of d-nucleotides and proteins made by ribosomal translation of l-amino acids. From the perspective of synthetic biology, however, particularly anabolic enzymes that could build the mirror-image counterparts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service