Skip to Content
Merck
All Photos(1)

Key Documents

474940

Sigma-Aldrich

5-Chloro-2-methylphenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NCS
CAS Number:
Molecular Weight:
183.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.646 (lit.)

bp

268-272 °C (lit.)

density

1.248 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Cl)cc1N=C=S

InChI

1S/C8H6ClNS/c1-6-2-3-7(9)4-8(6)10-5-11/h2-4H,1H3

InChI key

VDBDGAPNWNWUSA-UHFFFAOYSA-N

General description

5-Chloro-2-methylphenyl isothiocyanate is an isothiocyanate derivative. Its enthalpy of vaporization at boiling point (543.15K) has been reported to be 48.045kjoule/mol.

Application

5-Chloro-2-methylphenyl isothiocyanate may be used in the synthesis of N-(5-chloro-2-methyphenyl)-N′-(1-methylethyl)-thiourea (HDL376).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)
Thomas J F Nieland et al.
Journal of lipid research, 48(8), 1832-1845 (2007-05-30)
Treatment of atherosclerotic disease often focuses on reducing plasma LDL-cholesterol or increasing plasma HDL-cholesterol. We examined in vitro the effects on HDL receptor [scavenger receptor class B type I (SR-BI)] activity of three classes of clinical and experimental plasma HDL-cholesterol-elevating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service