47015
1-Fluoro-4-hydroxy-1,4-diazoniabicyclo[2,2,2]octane bis(tetrafluoroborate) on aluminum oxide
≥95.0% (T)
Synonym(s):
Accufluor™, Accufluor™ NFTh, Accufluor NFTh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95.0% (T)
form
solid
SMILES string
F[B-](F)(F)F.F[B-](F)(F)F.O[N+]12CC[N+](F)(CC1)CC2
InChI
1S/C6H13FN2O.2BF4/c7-8-1-4-9(10,5-2-8)6-3-8;2*2-1(3,4)5/h10H,1-6H2;;/q+2;2*-1
InChI key
DRMMNFGYCLJZKD-UHFFFAOYSA-N
Other Notes
Selective fluorinating agent
Legal Information
Accufluor is a trademark of Honeywell International Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron Letters, 36, 6769-6769 (1995)
Synlett, 693-693 (1996)
Development of GSKDevelopment of GSK's reagent guides ? embedding sustainability into reagent selections reagent guides ? embedding sustainability into reagent selection.
Green Chemistry, 15, 1542-1549 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service