All Photos(1)
About This Item
Linear Formula:
[C10H5(Br)OH]2
CAS Number:
Molecular Weight:
444.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
mp
204-206 °C (lit.)
SMILES string
Oc1ccc2cc(Br)ccc2c1-c3c(O)ccc4cc(Br)ccc34
InChI
1S/C20H12Br2O2/c21-13-3-5-15-11(9-13)1-7-17(23)19(15)20-16-6-4-14(22)10-12(16)2-8-18(20)24/h1-10,23-24H
InChI key
OORIFUHRGQKYEV-UHFFFAOYSA-N
General description
6,6′-Dibromo-1,1′-bi-2-naphthol is a 1,1′-bi-2-naphthol derivative. Vibrational circular dichroism (VCD), electronic circular dichroism (ECD) and optical rotatory dispersion (ORD) spectra for the enantiomers of 6,6′-dibromo-1,1′-bi-2-naphthol have been evaluated.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prasad L Polavarapu et al.
The journal of physical chemistry. A, 113(18), 5423-5431 (2009-04-16)
The experimental vibrational circular dichroism (VCD), electronic circular dichroism (ECD), and optical rotatory dispersion (ORD) spectra for both enantiomers of 6,6'-dibromo-1,1'-bi-2-naphthol have been measured. The corresponding quantum chemical predictions for three different orientations of hydroxyl groups in this molecule were
Diastereoselective and enantioselective glyoxylate-ene reaction catalyzed by new class of binaphthol-derived titanium complex.
Terada M, et al.
Tetrahedron Letters, 35(36), 6693-6696 (1996)
S Kobayashi et al.
Chirality, 12(5-6), 540-543 (2000-05-29)
A novel binuclear chiral zirconium catalyst was successfully used in enantioselective Strecker reactions. The catalyst was readily prepared from zirconium t-butoxide (Zr(OtBu)4), (R)-6,6'-dibromo-1, 1'-bi-2-naphthol ((R)-6-Br-BINOL), and (R)-3,3'-dibromo-1, 1'-bi-2-naphthol ((R)-3-Br-BINOL) to form unique binuclear structure. It was revealed that a combination
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service