Skip to Content
Merck
All Photos(1)

Key Documents

46052

Sigma-Aldrich

2,2,2-Trichloroethyl acetate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CCl3
CAS Number:
Molecular Weight:
191.44
Beilstein:
1757888
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

refractive index

n20/D 1.457

density

1.401 g/mL at 20 °C (lit.)

SMILES string

CC(=O)OCC(Cl)(Cl)Cl

InChI

1S/C4H5Cl3O2/c1-3(8)9-2-4(5,6)7/h2H2,1H3

InChI key

XHAXVDWUMCHTCY-UHFFFAOYSA-N

General description

2,2,2-Trichloroethyl acetate is used as acetyl donor in anhydrous, enzyme-catalyzed enantioselective acetylations.[1][2][3][4]

Application

2,2,2-Trichloroethyl acetate may be used in the enzyme catalyzed synthesis of enantiomerically pure prostaglandin intermediate, (1S,4R)-(-)-4-hydroxy-2-cyclopentenyl acetate.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.A. Djadchenko et al.
Journal of the Chemical Society. Perkin Transactions 1, 2001-2001 (1989)
Synthesis of (1S, 4R)-(-)-4-Hydroxy-2-cyclopentenyl Acetate by a Highly Enantioselective Enzyme-Catalyzed Transesterification in Organic Solvents
Theil F, et al.
Synthesis, 07, 540-541 (1988)
F. Theil et al.
Synthesis, 540-540 (1988)
G. Carrea et al.
Journal of the Chemical Society. Perkin Transactions 1, 1057-1057 (1989)
M. Therisod et al.
Journal of the American Chemical Society, 108, 5638-5638 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service