Skip to Content
Merck
All Photos(1)

Key Documents

457965

Sigma-Aldrich

Europium

chip, in mineral oil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Eu
CAS Number:
Molecular Weight:
151.96
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace rare earth metals basis

form

chips in mineral oil

reaction suitability

reagent type: catalyst
core: europium

resistivity

81 μΩ-cm, 20°C

bp

1527 °C (lit.)

mp

822 °C (lit.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

[Eu]

InChI

1S/Eu

InChI key

OGPBJKLSAFTDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Rodriguez-Liviano et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3411-3418 (2013-02-14)
A procedure for the synthesis of multifunctional europium(III)-doped gadolinium(III) fluoride (Eu:GdF3) nanoparticles (~85 nm) with quasispherical shape by precipitation at 120 °C from diethylene glycol solutions containing lanthanide chlorides and an ionic liquid (1-Butyl, 2-methylimidazolium tetrafluoroborate) as fluoride source has
Guihua Cui et al.
Carbohydrate polymers, 94(1), 77-81 (2013-04-03)
A series of thermo-responsive copolymers of poly(N-isopropylacrylamide) (PNIPAM) and cellulose were synthesized via atom transfer radical polymerization (ATRP) using N-isopropylacrylamide as the monomer, cellulose acetate as the initiator, and CuCl/tris(2-dimethylaminoethyl)amine (Me6TREN) as a catalytic system. The resulting polymers had a
Min Zheng et al.
ACS applied materials & interfaces, 5(3), 1078-1083 (2013-01-22)
Iron is one of the most important elements in the metabolic process for all living system. However, both its deficiency and excess from normal permissible limits can induce serious disorders. We synthesized a europium-based metal-organic framework (Eu-MOF), EuL(3) (L =
Mingjing Liu et al.
Analytica chimica acta, 761, 149-156 (2013-01-15)
Time-resolved luminescence bioassay technique using lanthanide complexes as luminescent probes/sensors has shown great utilities in clinical diagnostics and biotechnology discoveries. In this work, a novel terpyridine polyacid derivative that can form highly stable complexes with lanthanide ions in aqueous media
James W Walton et al.
Chemical communications (Cambridge, England), 49(16), 1600-1602 (2013-01-22)
The synthesis, structure and photophysical properties of a series of highly emissive europium complexes is reported. Certain complexes enter mammalian cells by macropinocytosis and stain the mitochondria selectively, allowing observation of the Eu emission in cellulo by time-gated spectral imaging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service