Skip to Content
Merck
All Photos(1)

Key Documents

446181

Sigma-Aldrich

Trichlorocyclopentylsilane

95%

Synonym(s):

(Trichlorosilyl)cyclopentane, Cyclopentyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9SiCl3
CAS Number:
Molecular Weight:
203.57
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

181 °C (lit.)

density

1.226 g/mL at 25 °C (lit.)

SMILES string

Cl[Si](Cl)(Cl)C1CCCC1

InChI

1S/C5H9Cl3Si/c6-9(7,8)5-3-1-2-4-5/h5H,1-4H2

InChI key

FCMZRNUHEXJWGB-UHFFFAOYSA-N

General description

Trichloro(cyclopentyl)silane is a polyhedral oligomeric silsesquioxane (POSS) derivate which is synthesized by hydrosilylation of cyclopentene and trichlorosilane.

Application

Trichloro(cyclopentyl)silane can be used in the preparation of POSS by hydrolysis and condensation reaction that is majorly used to enhance the thermo-mechanical properties of different polymers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silsesquioxanes as ligands for gold clusters.
Schmid G, et al.
European Journal of Inorganic Chemistry, 1998(6), 813-817 (1998)
Surface modification of polyehtersulfone hollow fiber membrane using layer-by-layer nanocoating method.
So J, et al.
Polymer (Korea), 89-90 (2015)
Surface modification of polyehtersulfone hollow fiber membrane using layer-by-layer nanocoating method.
So J, et al.
Polymer (Korea), 89-90 (2015)
Influence of symmetry/asymmetry of the nanoparticles structure on the thermal stability of polyhedral oligomeric silsesquioxane/polystyrene nanocomposites.
Blanco I, et al.
Polymer Composites, 33(11), 1903-1910 (2012)
Ignazio Blanco et al.
Molecules (Basel, Switzerland), 25(13) (2020-07-02)
Novel polystyrene (PS)/polyhedral oligomeric silsequioxanes (POSSs) nanocomposites were designed and prepared by in situ polymerization, using, for the first time, three-cage POSS molecules. The synthesized compounds were first characterized by Fourier transform infrared spectroscopy (FTIR) and 1H NMR spectroscopy to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service