Skip to Content
Merck
All Photos(1)

Key Documents

442550

Sigma-Aldrich

Tris[trinitratocerium(IV)] paraperiodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ce(NO3)3]3(H2IO6)
CAS Number:
Molecular Weight:
1203.31
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

reaction suitability

reagent type: catalyst
core: cerium

mp

90 °C (lit.)

SMILES string

[Ce+4].[Ce+4].[Ce+4].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.OI(O)([O-])([O-])([O-])=O

InChI

1S/3Ce.H5IO6.9NO3/c;;;2-1(3,4,5,6)7;9*2-1(3)4/h;;;(H5,2,3,4,5,6,7);;;;;;;;;/q3*+4;;9*-1/p-3

InChI key

UAFAVRCQSOGROV-UHFFFAOYSA-K

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service