Skip to Content
Merck
All Photos(2)

Documents

441686

Sigma-Aldrich

3-Methoxyphenylboronic acid

Synonym(s):

(m-Methoxyphenyl)boronic acid, 3-Methoxybenzeneboronic acid, m-Anisylboronic acid, m-Methoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96
Beilstein:
2831223
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

160-163 °C (lit.)

SMILES string

COc1cccc(c1)B(O)O

InChI

1S/C7H9BO3/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5,9-10H,1H3

InChI key

NLLGFYPSWCMUIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gözde Murat Saltan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 372-381 (2017-08-02)
In an approach to develop efficient organic optoelectronic devices to be used in light-driven systems, a series of three thiophene linked benzimidazole conjugates were synthesized and characterized. The combination of two thiophene rings to a benzimidazole core decorated with different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service