Skip to Content
Merck
All Photos(1)

Key Documents

429341

Sigma-Aldrich

Gallium(III) iodide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Gallium iodide, Gallium triiodide, Triiodogallium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaI3
CAS Number:
Molecular Weight:
450.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gallium

mp

345 °C (subl.) (lit.)

density

4.15 g/mL at 25 °C (lit.)

SMILES string

I[Ga](I)I

InChI

1S/Ga.3HI/h;3*1H/q+3;;;/p-3

InChI key

DWRNSCDYNYYYHT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service