Skip to Content
Merck
All Photos(1)

Key Documents

424137

Sigma-Aldrich

IR-786 iodide

Dye content 97 %

Synonym(s):

2-[2-[2-Chloro-3-[(1,3-dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,3,3-trimethylindolium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36ClIN2
CAS Number:
Molecular Weight:
611.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 97%

mp

270 °C (dec.) (lit.)

λmax

775 nm

SMILES string

[I-].CN1c2ccccc2C(C)(C)/C1=C\C=C3/CCCC(\C=C\C4=[N+](C)c5ccccc5C4(C)C)=C3Cl

Application

IR laser dye

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service