Skip to Content
Merck
All Photos(1)

Key Documents

421359

Sigma-Aldrich

MERPOL® SH surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€61.00

€61.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€61.00

About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€61.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

100 °C (lit.)

transition temp

cloud point 48 °C (2 wt. % aqueous solution)

density

0.96 g/mL at 25 °C (lit.)

HLB

12

Application

Nonionic, ethylene oxide condensate. General purpose detergent and wetting agent.

Legal Information

MERPOL is a registered trademark of Stepan Co.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.1 - 100.0 °F - closed cup

Flash Point(C)

35.6 - 37.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service