Skip to Content
Merck
All Photos(1)

Key Documents

417513

Sigma-Aldrich

6-Bromochromone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5BrO2
CAS Number:
Molecular Weight:
225.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

135-139 °C (dec.) (lit.)

SMILES string

Brc1ccc2OC=CC(=O)c2c1

InChI

1S/C9H5BrO2/c10-6-1-2-9-7(5-6)8(11)3-4-12-9/h1-5H

InChI key

XVNBWGGBXOJIDR-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of 6-bromochromone, C9H5BrO2.
Staples RJ and Lea W.
Zeitschrift fur Kristallographie, 220(3), 371-372 (2005)
Ferrocenyl derivatives of pterocarpene and coumestan: Synthesis, structure and anticancer activity studies.
Kowalski K, et al.
Journal of Organometallic Chemistry, 772, 49-59 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service