Skip to Content
Merck
All Photos(3)

Documents

410756

Sigma-Aldrich

Diethyl (2-oxo-2-phenylethyl)phosphonate

97%

Synonym(s):

Diethyl benzoylmethylphosphonate, Diethyl phenacylphosphonate, NSC 648426

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
256.23
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.513 (lit.)

bp

192-193 °C/11 mmHg (lit.)

density

1.179 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CC(=O)c1ccccc1)OCC

InChI

1S/C12H17O4P/c1-3-15-17(14,16-4-2)10-12(13)11-8-6-5-7-9-11/h5-9H,3-4,10H2,1-2H3

InChI key

HPEVTTNSIPGLEL-UHFFFAOYSA-N

Application

Reactant involved in:
  • Asymmetric Michael addition of β-oxo phosphonates to nitro olefins
  • Gem-chlorofluorination of keto phosphonates with subsequent functionalization of the products
  • Cyclocondensation reactions to produce arylphosphonates
  • Diazo transfer reactions for synthesis of diazo-phosphonyl compounds
  • Horner-Wadsworth-Emmons reactions
  • Inverse-electron-demand Diels-Alder reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglass F Taber et al.
Tetrahedron letters, 49(48), 6904-6906 (2009-12-01)
An convenient reagent for the one-carbon homologation of an aldehyde to the corresponding alkyne is reported. This reagent allows this conversion to conveniently be carried out on a large scale under ambient conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service